Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199462.1 | NP_001186391.1 | 7 | 965 | ||
NM_002598.3 | NP_002589.2 | 6 | 1102 | |||
GenBank mRNA | AK055180.1 | - | 2 | 979 | ||
AY948416.1 | - | 7 | 892 | |||
BC008378.1 | - | 6 | 1064 | 4102043 |