Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGXB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003681.2 | NP_001003681.1 | 5 | 597 | ||
NR_027780.1 | 6 | 712 | ||||
XM_006724100.3 | XP_006724163.1 | 4 | 676 | |||
XM_006724101.3 | XP_006724164.1 | 4 | 676 | |||
XM_006724102.1 | XP_006724165.1 | 5 | 580 | |||
GenBank mRNA | AJ010070.1 | - | 6 | 661 | ||
AK299833.1 | - | 6 | 677 | |||
AL079310.1 | - | 6 | 661 | |||
BC004194.2 | - | 3 | 312 | |||
BC017326.2 | - | 5 | 549 | |||
BC025303.2 | - | 4 | 492 | |||
BC052624.1 | - | 5 | 551 | |||
BC062413.1 | - | 5 | 549 | |||
BC080611.1 | - | 6 | 667 | |||
BC104754.1 | - | 1 | 159 | 8143757 | ||
BC104756.1 | - | 1 | 159 | 8143759 | ||
BC111373.1 | - | 4 | 501 | |||
CR456504.1 | - | 4 | 453 |