Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129884.2 | NP_001123356.1 | 5 | 769 | ||
NM_001243725.1 | NP_001230654.1 | 4 | 877 | |||
NM_001243726.1 | NP_001230655.1 | 4 | 661 | |||
NM_001243727.2 | NP_001230656.1 | 3 | 495 | |||
NM_001278195.1 | NP_001265124.1 | 5 | 809 | |||
NM_015926.5 | NP_057010.1 | 4 | 717 | |||
NR_024012.2 | 3 | 425 | ||||
NR_103462.1 | 3 | 388 | ||||
NR_103463.1 | 3 | 357 | ||||
XM_006713195.3 | XP_006713258.1 | 4 | 657 | |||
XM_006713197.3 | XP_006713260.1 | 4 | 624 | |||
XM_006713198.2 | XP_006713261.1 | 4 | 692 | |||
XM_011533805.2 | XP_011532107.1 | 4 | 1372 | |||
XM_017006573.1 | XP_016862062.1 | 4 | 877 | |||
GenBank mRNA | AF072733.1 | - | 3 | 615 | ||
AK222938.1 | - | 4 | 629 | |||
AK222949.1 | - | 4 | 629 | |||
AL832608.1 | - | 2 | 3266 | |||
AY358972.1 | - | 3 | 616 | |||
BC001592.1 | - | 4 | 639 | |||
BC008742.2 | - | 4 | 877 | 3346733 | ||
BG326586.1 | - | 3 | 412 | |||
BG772477.1 | - | 3 | 430 | |||
CB142778.1 | - | 3 | 389 | |||
HY149215.1 | - | 3 | 411 |