Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB1I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042454.2 | NP_001035919.1 | 5 | 420 | ||
NM_001164719.1 | NP_001158191.1 | 5 | 389 | |||
NM_015927.4 | NP_057011.2 | 5 | 419 | |||
GenBank mRNA | AB007836.1 | - | 4 | 384 | ||
AF116343.1 | - | 4 | 335 | |||
AK122975.1 | - | 3 | 1743 | |||
AK313327.1 | - | 5 | 420 | |||
BC001507.3 | - | 5 | 419 | 3030477 | ||
BC001830.2 | - | 5 | 419 | 3030477 | ||
BC017288.2 | - | 5 | 389 | 4901903 | ||
BC032545.1 | - | 4 | 391 | 5535913 |