Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF077202.1 | - | 4 | 212 | ||
AF161448.1 | - | 3 | 161 | |||
AK126594.1 | - | 4 | 2072 | |||
AL712164.1 | - | 4 | 189 | |||
AY508979.1 | - | 1 | 195 | |||
AY794023.1 | - | 4 | 196 | |||
BC001102.2 | - | 1 | 184 | |||
BC080531.1 | - | 4 | 197 | 4875991 | ||
BU536461.1 | - | 4 | 241 |