Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 1, 2 | 501 | ||
NM_015935.4 | NP_057019.3 | 1, 2 | 501 | |||
XM_017001458.1 | XP_016856947.1 | 1, 2 | 618 | |||
GenBank mRNA | AB020666.2 | - | 1, 2 | 256 | ||
AB242174.1 | - | 1, 2 | 426 | |||
AB242175.1 | - | 1, 2 | 414 | |||
AF132936.1 | - | 1, 2 | 426 | |||
AK027552.1 | - | 1, 2 | 420 | |||
AK223493.1 | - | 1, 2 | 462 | |||
AK291740.1 | - | 1, 2 | 448 | |||
AK304471.1 | - | 2, 3 | 282 | |||
AL049669.1 | - | 1, 2 | 426 | |||
BC029083.1 | - | 1, 2 | 255 | 4509086 | ||
BI091265.1 | - | 1, 2 | 617 |