Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000458.3 | NP_000449.1 | 9 | 2594 | ||
NM_001165923.3 | NP_001159395.1 | 9 | 2516 | |||
NM_001304286.1 | NP_001291215.1 | 7 | 2202 | |||
XM_011525160.1 | XP_011523462.1 | 8 | 2456 | |||
XM_011525161.1 | XP_011523463.1 | 8 | 2380 | |||
XM_011525164.1 | XP_011523466.1 | 8 | 2378 | |||
GenBank mRNA | BC017714.1 | - | 9 | 2588 | 4414015 | |
X58840.1 | - | 9 | 2594 |