Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302769.1 | NP_001289698.1 | 12 | 1907 | ||
NM_057177.6 | NP_476518.4 | 12 | 1907 | |||
NM_152526.5 | NP_689739.4 | 11 | 1721 | |||
NM_205863.3 | NP_995585.2 | 12 | 1907 | |||
XM_011510552.2 | XP_011508854.1 | 15 | 2250 | |||
XM_011510553.2 | XP_011508855.1 | 14 | 6973 | |||
XM_017003283.1 | XP_016858772.1 | 11 | 2197 | |||
XM_017003284.1 | XP_016858773.1 | 12 | 5243 | |||
XM_017003285.1 | XP_016858774.1 | 13 | 6794 | |||
XM_017003286.1 | XP_016858775.1 | 11 | 5139 | |||
XM_017003287.1 | XP_016858776.1 | 14 | 6976 | |||
XM_017003288.1 | XP_016858777.1 | 14 | 6978 | |||
XM_017003289.1 | XP_016858778.1 | 9 | 1354 | |||
XM_017003290.1 | XP_016858779.1 | 13 | 1854 | |||
XM_017003291.1 | XP_016858780.1 | 12 | 1726 | |||
XM_017003292.1 | XP_016858781.1 | 12 | 1788 | |||
XM_017003293.1 | XP_016858782.1 | 9 | 1292 | |||
XM_017003294.1 | XP_016858783.1 | 14 | 6970 | |||
GenBank mRNA | AB073472.1 | - | 14 | 1700 | ||
AB092439.1 | - | 14 | 1700 | |||
AF428250.1 | - | 12 | 1700 | |||
AF428251.1 | - | 11 | 1514 | |||
AF466152.1 | - | 12 | 1700 | |||
AK056157.1 | - | 14 | 1788 | |||
AK057965.1 | - | 7 | 1065 |