Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302769.1 | NP_001289698.1 | 5, 6 | 804 | ||
NM_057177.6 | NP_476518.4 | 5, 6 | 804 | |||
NM_152526.5 | NP_689739.4 | 5, 6 | 804 | |||
NM_205863.3 | NP_995585.2 | 5, 6 | 804 | |||
XM_011510552.2 | XP_011508854.1 | 8, 9 | 1147 | |||
XM_011510553.2 | XP_011508855.1 | 7, 8 | 5870 | |||
XM_017003283.1 | XP_016858772.1 | 4, 5 | 1094 | |||
XM_017003284.1 | XP_016858773.1 | 5, 6 | 4140 | |||
XM_017003285.1 | XP_016858774.1 | 7, 8 | 5877 | |||
XM_017003286.1 | XP_016858775.1 | 4, 5 | 4036 | |||
XM_017003287.1 | XP_016858776.1 | 7, 8 | 5873 | |||
XM_017003288.1 | XP_016858777.1 | 7, 8 | 5875 | |||
XM_017003289.1 | XP_016858778.1 | 2, 3 | 251 | |||
XM_017003290.1 | XP_016858779.1 | 6, 7 | 751 | |||
XM_017003291.1 | XP_016858780.1 | 5, 6 | 623 | |||
XM_017003292.1 | XP_016858781.1 | 5, 6 | 685 | |||
XM_017003293.1 | XP_016858782.1 | 2, 3 | 189 | |||
XM_017003294.1 | XP_016858783.1 | 7, 8 | 5867 | |||
GenBank mRNA | AB073472.1 | - | 7, 8 | 597 | ||
AB092439.1 | - | 7, 8 | 597 | |||
AF428250.1 | - | 5, 6 | 597 | |||
AF428251.1 | - | 5, 6 | 597 | |||
AF466152.1 | - | 5, 6 | 597 | |||
AK056157.1 | - | 7, 8 | 686 |