Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290265.1 | NP_001277194.1 | 3, 4 | 508 | ||
NM_021267.4 | NP_067090.1 | 3, 4 | 662 | |||
NM_198207.2 | NP_937850.1 | 3, 4 | 662 | |||
GenBank mRNA | AK293463.1 | - | 3, 4 | 508 | ||
BC084582.1 | - | 3, 4 | 460 | 6192817 |
Gene Symbol: GDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001492.5 | NP_001483.3 | 3, 4 | 662 | ||
GenBank mRNA | M62302.1 | - | 3, 4 | 614 |