Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286272.1 | NP_001273201.1 | 6 | 991 | ||
NM_001286273.1 | NP_001273202.1 | 5 | 752 | |||
NM_003295.3 | NP_003286.1 | 6 | 826 | |||
GenBank mRNA | AK130847.1 | - | 6 | 725 | ||
AK296587.1 | - | 6 | 635 | |||
AY117678.1 | - | 6 | 726 | |||
BC003352.2 | - | 6 | 709 | 2899964 | ||
BC012431.1 | - | 6 | 685 | 3880559 | ||
BC022436.1 | - | 5 | 680 | 4281364 | ||
BC040008.1 | - | 3 | 1732 | |||
BC052333.1 | - | 6 | 715 | 6058799 | ||
L13806.1 | - | 4 | 446 | |||
X16064.1 | - | 6 | 705 | |||
X64899.1 | - | 6 | 697 |