Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286272.1 | NP_001273201.1 | 6 | 907 | ||
NM_001286273.1 | NP_001273202.1 | 4, 5 | 668 | |||
NM_003295.3 | NP_003286.1 | 5, 6 | 742 | |||
GenBank mRNA | AK130847.1 | - | 5, 6 | 641 | ||
AK296587.1 | - | 5, 6 | 551 | |||
AY117678.1 | - | 5, 6 | 642 | |||
BC003352.2 | - | 5, 6 | 625 | 2899964 | ||
BC012431.1 | - | 5, 6 | 601 | 3880559 | ||
BC022436.1 | - | 4, 5 | 596 | 4281364 | ||
BC040008.1 | - | 2, 3 | 1648 | |||
BC052333.1 | - | 5, 6 | 631 | 6058799 | ||
L13806.1 | - | 3, 4 | 362 | |||
X16064.1 | - | 5, 6 | 621 | |||
X64899.1 | - | 5, 6 | 613 |