Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173531.2 | NP_001167002.1 | 5 | 1527 | ||
NM_001285986.1 | NP_001272915.1 | 3 | 2238 | |||
NM_001285987.1 | NP_001272916.1 | 4 | 2013 | |||
NM_002701.5 | NP_002692.2 | 5 | 1368 | |||
NM_203289.5 | NP_976034.4 | 4 | 2013 | |||
GenBank mRNA | BC020712.1 | - | 4 | 1093 | ||
DQ486514.1 | - | 3 | 2237 | |||
DQ486515.1 | - | 4 | 1681 | |||
DQ486516.1 | - | 5 | 1195 | |||
DQ486517.1 | - | 5 | 1003 | |||
KF880691.1 | - | 5 | 1570 | |||
KF880692.1 | - | 3 | 2281 | |||
KF880693.1 | - | 4 | 1651 | |||
KJ624996.1 | - | 5 | 1325 | |||
KJ700311.1 | - | 4 | 1651 | |||
KM188057.1 | - | 4 | 1894 | |||
Z11898.1 | - | 5 | 1336 | |||
Z11899.1 | - | 4 | 1114 |
Gene Symbol: POU5F1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159542.1 | NP_001153014.1 | 1 | 1546 | ||
GenBank mRNA | DQ486513.1 | - | 1 | 1546 |