Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015267.3 | NP_056082.2 | 7 | 732 | ||
XM_011538061.2 | XP_011536363.1 | 7 | 2242 | |||
XM_011538063.2 | XP_011536365.1 | 6 | 2107 | |||
XM_011538069.2 | XP_011536371.1 | 7 | 581 | |||
XM_011538070.2 | XP_011536372.1 | 7 | 1491 | |||
XM_017019080.1 | XP_016874569.1 | 8 | 1574 | |||
XM_017019081.1 | XP_016874570.1 | 8 | 637 | |||
GenBank mRNA | AB006631.2 | - | 7 | 731 | ||
BC151245.1 | - | 7 | 742 | 8860519 |