Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292018.1 | NP_001278947.1 | 7, 8 | 1067 | ||
NM_001292019.1 | NP_001278948.1 | 7, 8 | 633 | |||
NM_004762.3 | NP_004753.1 | 7, 8 | 670 | |||
NM_017456.3 | NP_059430.2 | 7, 8 | 670 | |||
XM_011525475.2 | XP_011523777.1 | 6, 7 | 529 | |||
XM_011525476.2 | XP_011523778.1 | 6, 7 | 529 | |||
XM_011525477.1 | XP_011523779.1 | 5, 6 | 448 | |||
XM_011525478.1 | XP_011523780.1 | 6, 7 | 989 | |||
XM_017025336.1 | XP_016880825.1 | 7, 8 | 740 | |||
XM_017025337.1 | XP_016880826.1 | 7, 8 | 625 | |||
XM_017025338.1 | XP_016880827.1 | 7, 8 | 1056 | |||
GenBank mRNA | AK293894.1 | - | 7, 8 | 1067 | ||
AK296914.1 | - | 4, 5 | 195 | |||
AK307917.1 | - | 7, 8 | 599 | |||
AK316075.1 | - | 7, 8 | 626 | |||
AK316277.1 | - | 7, 8 | 614 | |||
BC038385.1 | - | 7, 8 | 591 | 4755203 | ||
BC050452.1 | - | 7, 8 | 580 | 6043039 | ||
BX339155.2 | - | 7, 8 | 650 | |||
M85169.1 | - | 7, 8 | 620 |