Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020884.4 | NP_065935.3 | 13 | 1263 | ||
XM_006723840.3 | XP_006723903.1 | 11 | 904 | |||
XM_011528941.2 | XP_011527243.1 | 10 | 868 | |||
XM_011528947.2 | XP_011527249.1 | 4 | 1241 | |||
XM_011528948.2 | XP_011527250.1 | 4 | 400 | |||
XM_017027986.1 | XP_016883475.1 | 11 | 904 | |||
XM_017027987.1 | XP_016883476.1 | 6 | 419 | |||
GenBank mRNA | AB040945.2 | - | 11 | 972 | ||
BC151242.1 | - | 11 | 984 | 8860515 |