siRNA Details
Gene Symbol: ATRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207047.2 | NP_001193976.1 | 16 | 2448 | ||
NM_001323332.1 | NP_001310261.1 | 16 | 2831 | |||
NM_139321.2 | NP_647537.1 | 16 | 2831 | |||
NM_139322.3 | NP_647538.1 | 16 | 2831 | |||
GenBank mRNA | AF106861.1 | - | 16 | 2741 | ||
AK000356.1 | - | 1 | 148 | |||
AK293010.1 | - | 16 | 2831 | |||
AK302730.1 | - | 16 | 2427 | |||
BC101705.1 | - | 16 | 2781 | 8069211 | ||
DA368237.1 | - | 2 | 352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
