Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008211.1 | NP_001008212.1 | 4 | 599 | ||
NM_001008212.1 | NP_001008213.1 | 3 | 530 | |||
NM_001008213.1 | NP_001008214.1 | 4 | 584 | |||
NM_021980.4 | NP_068815.2 | 2 | 378 | |||
GenBank mRNA | AF061034.1 | - | 4 | 472 | ||
AF420371.1 | - | 4 | 456 | |||
AF420372.1 | - | 2 | 235 | |||
AF420373.1 | - | 3 | 387 | |||
AK055403.1 | - | 3 | 819 | |||
AK303715.1 | - | 2 | 302 | |||
AK316171.1 | - | 2 | 324 | |||
BC013876.2 | - | 2 | 291 | 3831267 | ||
BC032762.1 | - | 4 | 504 | 3457195 | ||
KU178502.1 | - | 1 | 146 | |||
KU178503.1 | - | 1 | 146 | |||
KU178504.1 | - | 1 | 146 |