Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301076.1 | NP_001288005.1 | 2 | 215 | ||
NM_001301078.1 | NP_001288007.1 | 2 | 290 | |||
NM_001301081.1 | NP_001288010.1 | 2 | 172 | |||
NM_004069.4 | NP_004060.2 | 2 | 290 | |||
NM_021575.3 | NP_067586.1 | 2 | 290 | |||
XM_011526423.2 | XP_011524725.1 | 2 | 226 | |||
XM_011526424.2 | XP_011524726.1 | 2 | 121 | |||
GenBank mRNA | AJ010148.1 | - | 1 | 61 | ||
AJ010149.1 | - | 1 | 61 | |||
AJ713543.1 | - | 2 | 97 | |||
AK312003.1 | - | 2 | 142 | |||
BC006337.2 | - | 2 | 99 | 4304127 | ||
BG282225.1 | - | 2 | 98 | |||
BI561502.1 | - | 2 | 177 | |||
BQ221562.1 | - | 2 | 215 | |||
CA454481.1 | - | 2 | 205 | |||
CB108423.1 | - | 2 | 241 | |||
DB472500.1 | - | 2 | 290 | |||
H46780.1 | - | 2 | 262 | |||
X97074.1 | - | 2 | 175 |