Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020802.3 | NP_065853.3 | 6 | 1682 | ||
XM_005271627.4 | XP_005271684.2 | 6 | 1413 | |||
GenBank mRNA | AB037798.1 | - | 3 | 1025 | ||
AK095004.1 | - | 5 | 1312 | |||
AK296350.1 | - | 3 | 1423 | |||
BX647415.1 | - | 6 | 1432 |