Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005369.1 | NP_001005369.1 | 12 | 1964 | ||
NM_001321001.1 | NP_001307930.1 | 11 | 1499 | |||
NM_001321002.1 | NP_001307931.1 | 11 | 1471 | |||
NM_001321003.1 | NP_001307932.1 | 12 | 1681 | |||
NM_001321004.1 | NP_001307933.1 | 10 | 1733 | |||
NM_001321005.1 | NP_001307934.1 | 10 | 1382 | |||
NM_002453.2 | NP_002444.2 | 11 | 1543 | |||
XM_005264335.3 | XP_005264392.1 | 9 | 1342 | |||
XM_011532871.2 | XP_011531173.1 | 10 | 1376 | |||
XM_017004162.1 | XP_016859651.1 | 12 | 1672 | |||
XM_017004163.1 | XP_016859652.1 | 11 | 1748 | |||
XM_017004164.1 | XP_016859653.1 | 10 | 1360 | |||
XM_017004165.1 | XP_016859654.1 | 11 | 1575 | |||
XM_017004166.1 | XP_016859655.1 | 11 | 1767 | |||
XM_017004167.1 | XP_016859656.1 | 7 | 1844 | |||
GenBank mRNA | AF494407.1 | - | 11 | 1523 | ||
AK225184.1 | - | 11 | 1512 | |||
AK307993.1 | - | 11 | 1523 | |||
BC039848.1 | - | 11 | 1501 | |||
BC064977.1 | - | 11 | 1483 | |||
L34600.1 | - | 11 | 1497 |