Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005753.2 | NP_001005753.1 | 5 | 1602 | ||
NM_001193517.1 | NP_001180446.1 | 6 | 1543 | |||
NM_016079.3 | NP_057163.1 | 6 | 1663 | |||
NR_036454.1 | 4 | 1422 | ||||
GenBank mRNA | AF151907.1 | - | 6 | 1531 | ||
AK025046.1 | - | 1 | 559 | |||
AK025260.1 | - | 6 | 1561 | |||
AK129502.1 | - | 6 | 1420 | |||
BC004419.1 | - | 6 | 1561 | 3660116 |
Gene Symbol: RNF103-CHMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198954.1 | NP_001185883.1 | 8 | 1844 |