Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003150.3 | NP_003141.2 | 21 | 2273 | ||
NM_139276.2 | NP_644805.1 | 21 | 2295 | |||
NM_213662.1 | NP_998827.1 | 21 | 2187 | |||
XM_005257616.3 | XP_005257673.2 | 21 | 2286 | |||
XM_005257617.3 | XP_005257674.2 | 21 | 2281 | |||
XM_011525145.2 | XP_011523447.1 | 21 | 2286 | |||
XM_011525146.2 | XP_011523448.1 | 21 | 2286 | |||
XM_017024972.1 | XP_016880461.1 | 21 | 2281 | |||
XM_017024973.1 | XP_016880462.1 | 21 | 2286 | |||
XM_017024974.1 | XP_016880463.1 | 21 | 2281 | |||
XM_017024975.1 | XP_016880464.1 | 21 | 2274 | |||
XM_017024976.1 | XP_016880465.1 | 21 | 2274 | |||
GenBank mRNA | AB451232.1 | - | 20 | 2055 | ||
AF029311.1 | - | 3 | 367 | |||
AJ012463.1 | - | 20 | 2055 | |||
AK291933.1 | - | 21 | 2368 | |||
AK297994.1 | - | 20 | 2080 | |||
AK301200.1 | - | 21 | 2203 | |||
BC000627.2 | - | 21 | 2192 | 3347434 | ||
BC014482.1 | - | 21 | 2187 | 4909141 |