Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
Gene Name:
cAMP regulated phosphoprotein 21
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
UniGene ID:
Hs.475902
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
236375
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267617.1
NP_001254546.1
19
2733
NM_001267619.1
NP_001254548.1
19
2758
NM_016300.4
NP_057384.2
20
2807
XM_005264811.2
XP_005264868.1
21
2941
XM_006712943.2
XP_006713006.1
20
2839
XM_006712944.2
XP_006713007.1
19
2779
XM_011533298.2
XP_011531600.1
21
2944
XM_011533299.2
XP_011531601.1
21
3488
XM_011533300.2
XP_011531602.1
21
3107
XM_011533301.2
XP_011531603.1
21
2938
XM_011533302.2
XP_011531604.1
22
3030
XM_011533303.2
XP_011531605.1
22
3580
XM_017005574.1
XP_016861063.1
21
3034
XM_017005575.1
XP_016861064.1
22
14667
XM_017005576.1
XP_016861065.1
21
3485
XM_017005577.1
XP_016861066.1
21
2935
XM_017005578.1
XP_016861067.1
21
2911
XM_017005579.1
XP_016861068.1
21
3485
XM_017005580.1
XP_016861069.1
22
3577
XM_017005581.1
XP_016861070.1
22
14664
XM_017005582.1
XP_016861071.1
21
3482
XM_017005583.1
XP_016861072.1
21
2932
XM_017005584.1
XP_016861073.1
20
3428
XM_017005585.1
XP_016861074.1
20
3425
XM_017005586.1
XP_016861075.1
20
2881
XM_017005587.1
XP_016861076.1
20
3044
XM_017005588.1
XP_016861077.1
21
3517
XM_017005589.1
XP_016861078.1
21
2967
XM_017005590.1
XP_016861079.1
21
14604
XM_017005591.1
XP_016861080.1
20
3386
XM_017005592.1
XP_016861081.1
20
3383
XM_017005593.1
XP_016861082.1
20
2929
XM_017005594.1
XP_016861083.1
20
2833
XM_017005595.1
XP_016861084.1
21
3475
XM_017005596.1
XP_016861085.1
20
3383
XM_017005597.1
XP_016861086.1
20
3380
XM_017005598.1
XP_016861087.1
21
3472
XM_017005599.1
XP_016861088.1
19
3326
XM_017005600.1
XP_016861089.1
19
2945
XM_017005601.1
XP_016861090.1
19
2776
XM_017005602.1
XP_016861091.1
19
3323
XM_017005603.1
XP_016861092.1
19
2942
XM_017005604.1
XP_016861093.1
19
2773
XM_017005605.1
XP_016861094.1
20
3415
XM_017005606.1
XP_016861095.1
20
2865
XM_017005607.1
XP_016861096.1
20
3343
XM_017005608.1
XP_016861097.1
22
3576
XM_017005609.1
XP_016861098.1
22
3608
XM_017005610.1
XP_016861099.1
19
3274
XM_017005611.1
XP_016861100.1
18
3214
XM_017005612.1
XP_016861101.1
19
3271
GenBank mRNA
AK093648.1
-
1
1597
AK294484.1
-
19
2771
AL133109.1
-
13
2062
BC031106.1
-
8
1619
BC036399.1
-
19
2738
5263894
BC051828.1
-
20
2807
6012704
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0