Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
Gene Name:
cAMP regulated phosphoprotein 21
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
UniGene ID:
Hs.475902
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
236376
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267617.1
NP_001254546.1
17
2125
NM_001267619.1
NP_001254548.1
17
2150
NM_016300.4
NP_057384.2
18
2199
XM_005264811.2
XP_005264868.1
19
2333
XM_006712943.2
XP_006713006.1
18
2231
XM_006712944.2
XP_006713007.1
17
2171
XM_011533298.2
XP_011531600.1
19
2336
XM_011533299.2
XP_011531601.1
19
2880
XM_011533300.2
XP_011531602.1
19
2499
XM_011533301.2
XP_011531603.1
19
2330
XM_011533302.2
XP_011531604.1
20
2422
XM_011533303.2
XP_011531605.1
20
2972
XM_017005574.1
XP_016861063.1
19
2426
XM_017005575.1
XP_016861064.1
20
14059
XM_017005576.1
XP_016861065.1
19
2877
XM_017005577.1
XP_016861066.1
19
2327
XM_017005578.1
XP_016861067.1
19
2303
XM_017005579.1
XP_016861068.1
19
2877
XM_017005580.1
XP_016861069.1
20
2969
XM_017005581.1
XP_016861070.1
20
14056
XM_017005582.1
XP_016861071.1
19
2874
XM_017005583.1
XP_016861072.1
19
2324
XM_017005584.1
XP_016861073.1
18
2820
XM_017005585.1
XP_016861074.1
18
2817
XM_017005586.1
XP_016861075.1
18
2273
XM_017005587.1
XP_016861076.1
18
2436
XM_017005588.1
XP_016861077.1
19
2909
XM_017005589.1
XP_016861078.1
19
2359
XM_017005590.1
XP_016861079.1
19
13996
XM_017005591.1
XP_016861080.1
18
2778
XM_017005592.1
XP_016861081.1
18
2775
XM_017005593.1
XP_016861082.1
18
2321
XM_017005594.1
XP_016861083.1
18
2225
XM_017005595.1
XP_016861084.1
19
2867
XM_017005596.1
XP_016861085.1
18
2775
XM_017005597.1
XP_016861086.1
18
2772
XM_017005598.1
XP_016861087.1
19
2864
XM_017005599.1
XP_016861088.1
17
2718
XM_017005600.1
XP_016861089.1
17
2337
XM_017005601.1
XP_016861090.1
17
2168
XM_017005602.1
XP_016861091.1
17
2715
XM_017005603.1
XP_016861092.1
17
2334
XM_017005604.1
XP_016861093.1
17
2165
XM_017005605.1
XP_016861094.1
18
2807
XM_017005606.1
XP_016861095.1
18
2257
XM_017005607.1
XP_016861096.1
18
2735
XM_017005608.1
XP_016861097.1
20
2968
XM_017005609.1
XP_016861098.1
20
3000
XM_017005610.1
XP_016861099.1
17
2666
XM_017005611.1
XP_016861100.1
16
2606
XM_017005612.1
XP_016861101.1
17
2663
GenBank mRNA
AK294484.1
-
17
2163
AL133109.1
-
11
1454
BC031106.1
-
6
1011
BC036399.1
-
17
2130
5263894
BC051828.1
-
18
2199
6012704
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0