Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308165.1 | NP_001295094.1 | 4 | 488 | ||
NM_007017.2 | NP_008948.1 | 3 | 1591 | |||
NM_178424.1 | NP_848511.1 | 3 | 1591 | |||
XM_005265803.4 | XP_005265860.1 | 4 | 355 | |||
XM_011534420.1 | XP_011532722.1 | 4 | 1381 | |||
GenBank mRNA | AB022083.1 | - | 3 | 1544 | ||
AB022441.1 | - | 3 | 1544 | |||
AK302162.1 | - | 4 | 488 | |||
AK308329.1 | - | 3 | 1539 | |||
BC033492.1 | - | 3 | 1591 | 5172577 |