Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CNTN4 SIRNA Assays ›
Gene Symbol:
CNTN4
Gene Name:
contactin 4
Gene Aliases:
AXCAM, BIG-2
Chromosome Location:
Chr.3: 2098771 - 3059080 on Build GRCh38
UniGene ID:
Hs.298705
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
236481
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CNTN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001206955.1
NP_001193884.1
14
1952
NM_001206956.1
NP_001193885.1
6
674
NM_175607.2
NP_783200.1
15
1907
NM_175613.2
NP_783302.1
6
674
XM_006713004.3
XP_006713067.1
14
1920
XM_011533425.2
XP_011531727.1
14
1920
XM_011533426.2
XP_011531728.1
15
2139
XM_011533427.2
XP_011531729.1
15
2120
XM_011533428.2
XP_011531730.1
13
2109
XM_011533429.2
XP_011531731.1
13
2044
XM_011533430.2
XP_011531732.1
13
2005
XM_011533431.2
XP_011531733.1
12
4754
XM_017005782.1
XP_016861271.1
15
1862
XM_017005783.1
XP_016861272.1
16
2736
XM_017005784.1
XP_016861273.1
14
2197
XM_017005785.1
XP_016861274.1
15
1847
XM_017005786.1
XP_016861275.1
13
2109
XM_017005787.1
XP_016861276.1
13
2005
XM_017005788.1
XP_016861277.1
13
1718
GenBank mRNA
AF549455.1
-
12
1950
AK057278.1
-
6
652
AK095729.1
-
13
1765
AK097633.1
-
9
1198
AK125460.1
-
14
2098
AK314396.1
-
6
652
AY090737.1
-
14
1789
BC026119.1
-
6
657
4826091
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
Function(s)
neuron cell-cell adhesion
nervous system development
axonogenesis
axon guidance
axonal fasciculation
brain development
neuron projection development
negative regulation of neuron differentiation
regulation of synaptic plasticity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0