Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242466.1 | NP_001229395.1 | 9 | 4102 | ||
NM_181504.3 | NP_852556.2 | 10 | 4239 | |||
NM_181523.2 | NP_852664.1 | 16 | 5539 | |||
NM_181524.1 | NP_852665.1 | 10 | 4371 | |||
XM_005248542.3 | XP_005248599.1 | 16 | 5562 | |||
XM_011543493.2 | XP_011541795.1 | 15 | 5192 | |||
XM_017009585.1 | XP_016865074.1 | 16 | 6331 | |||
XM_017009586.1 | XP_016865075.1 | 15 | 5152 | |||
GenBank mRNA | BE888150.1 | - | 1 | 254 | ||
BQ723333.1 | - | 1 | 697 |