Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005103.4 | NP_005094.1 | 7, 8 | 1249 | ||
XM_005271734.2 | XP_005271791.1 | 7, 8 | 1294 | |||
XM_005271735.2 | XP_005271792.1 | 7, 8 | 1346 | |||
GenBank mRNA | BC009545.1 | - | 7, 8 | 1166 | 3897353 | |
CR456817.1 | - | 6, 7 | 1014 | |||
CR541954.1 | - | 6, 7 | 1014 | |||
U60060.1 | - | 7, 8 | 1113 | |||
U69139.1 | - | 6, 7 | 1039 |