Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005103.4 | NP_005094.1 | 10 | 1588 | ||
XM_005271734.2 | XP_005271791.1 | 10 | 1633 | |||
XM_005271735.2 | XP_005271792.1 | 10 | 1685 | |||
GenBank mRNA | BC009545.1 | - | 10 | 1505 | 3897353 | |
U60060.1 | - | 10 | 1452 | |||
U69139.1 | - | 9 | 1377 |