Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS27L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015920.3 | NP_057004.1 | 4 | 516 | ||
GenBank mRNA | AF070668.1 | - | 4 | 432 | ||
AK024591.1 | - | 4 | 445 | |||
BC003667.1 | - | 4 | 448 | 3827362 |