Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006624.1 | NP_001006625.1 | 5 | 551 | ||
NM_001006625.1 | NP_001006626.1 | 5 | 551 | |||
NM_006474.4 | NP_006465.3 | 5 | 727 | |||
NM_198389.2 | NP_938203.2 | 5 | 727 | |||
XM_006710295.1 | XP_006710358.1 | 5 | 458 | |||
GenBank mRNA | AY194238.1 | - | 5 | 653 |