Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000686.1 | XP_016856175.1 | 5 | 2004 | ||
GenBank mRNA | AB023193.1 | - | 5 | 2035 |