Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFASC SIRNA Assays ›
Gene Symbol:
NFASC
Gene Name:
neurofascin
Gene Aliases:
NF, NRCAML
Chromosome Location:
Chr.1: 204828652 - 205022822 on Build GRCh38
UniGene ID:
Hs.13349
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
236875
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFASC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005388.2
NP_001005388.2
30
4836
NM_001160331.1
NP_001153803.1
27
4445
NM_001160332.1
NP_001153804.1
28
4638
NM_015090.3
NP_055905.2
27
4623
XM_005244989.3
XP_005245046.2
29
4763
XM_005244991.3
XP_005245048.2
28
4718
XM_005244992.4
XP_005245049.2
27
4703
XM_005244993.3
XP_005245050.2
27
4667
XM_011509311.1
XP_011507613.1
30
4955
XM_011509312.1
XP_011507614.1
32
5270
XM_011509313.1
XP_011507615.1
30
4925
XM_011509314.1
XP_011507616.1
31
5219
XM_011509315.1
XP_011507617.1
29
4874
XM_011509316.1
XP_011507618.1
33
5167
XM_011509317.1
XP_011507619.1
30
4817
XM_011509318.2
XP_011507620.1
32
5192
XM_011509319.1
XP_011507621.1
32
5116
XM_011509320.2
XP_011507622.1
31
5141
XM_011509321.1
XP_011507623.1
30
4733
XM_011509322.1
XP_011507624.1
29
4700
XM_011509323.1
XP_011507625.1
29
4649
XM_011509324.1
XP_011507626.1
28
4463
XM_011509325.1
XP_011507627.1
27
4448
XM_011509326.1
XP_011507628.1
26
4142
XM_011509327.1
XP_011507629.1
25
4127
XM_011509328.1
XP_011507630.1
25
4091
XM_017000733.1
XP_016856222.1
31
5225
XM_017000734.1
XP_016856223.1
30
5174
XM_017000735.1
XP_016856224.1
29
4718
XM_017000736.1
XP_016856225.1
27
4418
XM_017000738.1
XP_016856227.1
28
4640
XM_017000739.1
XP_016856228.1
27
4625
XM_017000740.1
XP_016856229.1
25
4304
XM_017000741.1
XP_016856230.1
29
4760
XM_017000742.1
XP_016856231.1
28
4585
GenBank mRNA
AB018299.3
-
26
4454
AK027553.1
-
6
1578
AK127424.1
-
8
1918
AK128699.1
-
6
2412
AK172728.1
-
2
787
BX537841.1
-
4
1442
CR749402.1
-
3
2331
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein localization to paranode region of axon
axon guidance
peripheral nervous system development
transmission of nerve impulse
paranodal junction assembly
heterotypic cell-cell adhesion
myelination
clustering of voltage-gated sodium channels
synapse organization
protein localization to juxtaparanode region of axon
protein targeting to plasma membrane
protein binding
protein domain specific binding
protein binding involved in heterotypic cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0