Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015980.4 | NP_057064.1 | 5 | 2139 | ||
GenBank mRNA | AF113537.1 | - | 5 | 2005 | ||
AF133422.1 | - | 5 | 1982 | |||
AK098398.1 | - | 2547 | ||||
BC002619.2 | - | 5 | 2022 | 3160345 | ||
BC018873.2 | - | 5 | 2015 |