Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139498.1 | NP_001132970.1 | 6 | 1968 | ||
NM_001139500.1 | NP_001132972.1 | 7 | 2136 | |||
NM_001139501.1 | NP_001132973.1 | 8 | 2246 | |||
NM_001139502.1 | NP_001132974.1 | 6 | 1889 | |||
NM_004114.3 | NP_004105.1 | 5 | 2501 | |||
NM_033642.2 | NP_378668.1 | 5 | 1764 | |||
XM_005262399.1 | XP_005262456.1 | 7 | 2339 | |||
GenBank mRNA | AF100143.1 | - | 5 | 2068 | ||
AF100144.1 | - | 5 | 1763 | |||
BC012347.1 | - | 5 | 2498 | 4551396 | ||
BC034340.1 | - | 5 | 2204 | 3918908 |