Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040105.1 | NP_001035194.1 | 7 | 12914 | ||
GenBank mRNA | AF430017.1 | - | 5 | 2471 | ||
AJ606307.1 | - | 7 | 12914 | |||
AK026404.1 | - | 5 | 1585 | |||
BC126315.1 | - | 5 | 3056 |