Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077180.2 | NP_001070648.1 | 5 | 2472 | ||
NM_001288659.1 | NP_001275588.1 | 5 | 2132 | |||
NM_001288660.1 | NP_001275589.1 | 5 | 2129 | |||
NM_016025.4 | NP_057109.3 | 5 | 2475 | |||
XM_017023269.1 | XP_016878758.1 | 4 | 2168 | |||
GenBank mRNA | BC000195.1 | - | 5 | 2360 | 3350598 | |
BX647201.1 | - | 1 | 4822 |