Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252650.1 | NP_001239579.1 | 6 | 923 | ||
NM_016026.3 | NP_057110.3 | 7 | 1133 | |||
GenBank mRNA | AB209223.1 | - | 4 | 3194 | ||
AF151840.1 | - | 7 | 1082 | |||
AF167438.1 | - | 7 | 1083 | |||
AK057195.1 | - | 7 | 969 | |||
AK074749.1 | - | 7 | 1113 | |||
AK289427.1 | - | 7 | 1031 | |||
AK293355.1 | - | 6 | 863 | |||
BC000112.1 | - | 7 | 1050 | 3504920 | ||
BC011727.2 | - | 7 | 1048 | 4135182 | ||
BC026274.1 | - | 7 | 1070 | 4250934 | ||
BC037302.1 | - | 7 | 1110 | 5261204 | ||
BC051291.1 | - | 7 | 1043 | 3533223 |