Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305942.1 | NP_001292871.1 | 4 | 1229 | ||
NM_016035.4 | NP_057119.2 | 7 | 1550 | |||
GenBank mRNA | AF151850.1 | - | 7 | 1521 | ||
AK025376.1 | - | 7 | 1464 | |||
AK095083.1 | - | 3 | 3015 | |||
AK128853.1 | - | 2 | 5214 | |||
BC011895.2 | - | 7 | 1220 | 3944239 | ||
EU216419.1 | - | 7 | 1532 | |||
EU216420.1 | - | 7 | 1450 | |||
EU216421.1 | - | 7 | 1284 | |||
EU216422.1 | - | 7 | 1222 | |||
EU216423.1 | - | 6 | 1146 |