Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001994.2 | NP_001001994.1 | 4 | 751 | ||
NM_001001995.2 | NP_001001995.1 | 5 | 1069 | |||
NM_001001996.2 | NP_001001996.1 | 5 | 1069 | |||
NM_001318729.1 | NP_001305658.1 | 4 | 751 | |||
NM_005278.4 | NP_005269.1 | 4 | 949 | |||
XM_005274489.4 | XP_005274546.1 | 4 | 948 | |||
XM_011545497.2 | XP_011543799.1 | 5 | 760 | |||
XM_017029432.1 | XP_016884921.1 | 5 | 759 | |||
XM_017029433.1 | XP_016884922.1 | 4 | 453 | |||
XM_017029434.1 | XP_016884923.1 | 4 | 476 | |||
GenBank mRNA | AB209525.1 | - | 4 | 470 | ||
AK095657.1 | - | 5 | 823 | |||
AK294314.1 | - | 4 | 522 | |||
AK299675.1 | - | 6 | 817 | |||
BC008151.1 | - | 4 | 575 | 4215416 | ||
BC047295.1 | - | 5 | 779 | 4215095 | ||
U45955.1 | - | 4 | 549 |