Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016060.2 | NP_057144.1 | 4 | 372 | ||
XM_017024710.1 | XP_016880199.1 | 3 | 445 | |||
GenBank mRNA | AF151883.1 | - | 4 | 344 | ||
AK130946.1 | - | 3 | 275 | |||
AK311875.1 | - | 4 | 341 | |||
BC012539.1 | - | 4 | 347 | 4248123 |