Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014245.4 | NP_055060.1 | 2 | 329 | ||
NM_183237.2 | NP_899060.1 | 2 | 322 | |||
NR_037702.1 | 3 | 593 | ||||
NR_037703.1 | 3 | 586 | ||||
GenBank mRNA | AF092878.1 | - | 2 | 196 | ||
AF142060.1 | - | 2 | 196 | |||
AF164679.1 | - | 2 | 249 | |||
AF312226.1 | - | 3 | 460 | |||
AK311982.1 | - | 2 | 261 | |||
BC005966.1 | - | 2 | 222 | 4069078 | ||
BC008627.1 | - | 2 | 232 | 4177613 | ||
BG715867.1 | - | 2 | 268 | |||
BG716023.1 | - | 2 | 268 | |||
BG720797.1 | - | 3 | 481 | |||
BG742338.1 | - | 2 | 329 | |||
BT007348.1 | - | 2 | 196 |