Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201370.1 | NP_001188299.1 | 2 | 425 | ||
NM_014245.4 | NP_055060.1 | 3 | 473 | |||
NM_183237.2 | NP_899060.1 | 3 | 466 | |||
NR_037702.1 | 4 | 737 | ||||
NR_037703.1 | 4 | 730 | ||||
GenBank mRNA | AF092878.1 | - | 3 | 340 | ||
AF164679.1 | - | 3 | 393 | |||
AF312226.1 | - | 4 | 604 | |||
BC005966.1 | - | 3 | 366 | 4069078 | ||
BC008627.1 | - | 3 | 376 | 4177613 | ||
BG715867.1 | - | 3 | 412 | |||
BG716023.1 | - | 3 | 412 | |||
BG720797.1 | - | 4 | 625 | |||
BG742338.1 | - | 3 | 473 | |||
DB272382.1 | - | 2 | 357 |