Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318970.1 | NP_001305899.1 | 6 | 561 | ||
NM_001623.4 | NP_001614.3 | 6 | 622 | |||
NM_032955.2 | NP_116573.1 | 3 | 451 | |||
GenBank mRNA | AF088038.1 | - | 6 | 568 | ||
AV716806.1 | - | 6 | 542 | |||
BC009474.1 | - | 6 | 601 | 4249306 | ||
CD249922.1 | - | 3 | 487 | |||
CD521003.1 | - | 6 | 529 | |||
U49392.1 | - | 6 | 580 |