Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006756.3 | NP_006747.1 | 10 | 2627 | ||
NM_201437.2 | NP_958845.1 | 9 | 2564 | |||
NR_109901.1 | 12 | 3003 | ||||
NR_109902.1 | 5 | 2075 | ||||
XM_006716467.1 | XP_006716530.1 | 11 | 2530 | |||
XM_011517578.1 | XP_011515880.1 | 10 | 2558 | |||
GenBank mRNA | BC072460.1 | - | 10 | 2423 | 6470030 | |
BX538034.1 | - | 12 | 2814 | |||
X57198.1 | - | 9 | 2357 | |||
X62585.1 | - | 10 | 2400 |