Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193517.1 | NP_001180446.1 | 2 | 232 | ||
NM_016079.3 | NP_057163.1 | 2 | 232 | |||
GenBank mRNA | AF151907.1 | - | 2 | 101 | ||
AF219226.1 | - | 2 | 83 | |||
AK025260.1 | - | 2 | 130 | |||
AK129502.1 | - | 2 | 109 | |||
AK312353.1 | - | 2 | 178 | |||
AY364249.1 | - | 2 | 83 | |||
BC004419.1 | - | 2 | 130 | 3660116 | ||
BG705423.1 | - | 2 | 219 | |||
BI562428.1 | - | 2 | 220 | |||
DA290886.1 | - | 2 | 232 |
Gene Symbol: RNF103-CHMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198954.1 | NP_001185883.1 | 4 | 413 | ||
GenBank mRNA | AK294389.1 | - | 4 | 413 |