Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014931.3 | NP_055746.3 | 24 | 3434 | ||
GenBank mRNA | AB029038.2 | - | 24 | 3759 | ||
AK126810.1 | - | 7 | 2186 | |||
BC002799.2 | - | 23 | 3036 | |||
BC007629.1 | - | 8 | 1280 | |||
BC068014.1 | - | 23 | 3098 | |||
BC094753.1 | - | 24 | 3296 | 4838365 |