Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000612.5 | NP_000603.1 | 4 | 4734 | ||
NM_001007139.5 | NP_001007140.2 | 5 | 4708 | |||
NM_001127598.2 | NP_001121070.1 | 5 | 4421 | |||
NM_001291861.2 | NP_001278790.1 | 4 | 4085 | |||
NM_001291862.2 | NP_001278791.1 | 4 | 4256 | |||
GenBank mRNA | AA040827.1 | - | 1 | 35 | ||
AF318382.1 | - | 1 | 2185 | |||
AK025719.1 | - | 1 | 1854 | |||
AY971350.1 | - | 1 | 235 | |||
BC011786.2 | - | 1 | 252 | |||
BC042127.1 | - | 2 | 477 |
Gene Symbol: INS-IGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003512.3 | 7 | 4696 |