Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001234.4 | NP_001225.1 | 1 | 121 | ||
NM_033337.2 | NP_203123.1 | 1 | 121 | |||
GenBank mRNA | AF036365.1 | - | 1 | 91 | ||
AF043101.1 | - | 1 | 41 | |||
AK291892.1 | - | 1 | 121 | |||
BC069368.1 | - | 1 | 94 | 7262168 | ||
BC102033.1 | - | 1 | 81 | 40033505 | ||
BC102036.1 | - | 1 | 81 | 40033114 | ||
BC102037.1 | - | 1 | 81 | 40033118 | ||
Y14747.1 | - | 1 | 55 |